Learn More
mitogen-activated protein kinase-activated protein kinase 2, Mouse, Clone: 3B8, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a member of the Ser/Thr protein kinase family. This kinase is regulated through direct phosphorylation by p38 MAP kinase. In conjunction with p38 MAP kinase, this kinase is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation and cell proliferation. Heat shock protein HSP27 was shown to be one of the substrates of this kinase in vivo. Two transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | mitogen-activated protein kinase-activated protein kinase 2 |
| Applications | ELISA, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | 3B8 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant MAPKAPK2. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | MAPKAPK2 |
| Gene Accession No. | NM_032960 |
| Gene Alias | MK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.