Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mas Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
$204.00 - $482.50
Specifications
Antigen | Mas |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:25, Immunohistochemistry-Paraffin 1:100 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP178444SS
|
Novus Biologicals
NBP178444SS |
0.025 mL |
Each for $204.00
|
|
|||||
NBP178444
|
Novus Biologicals
NBP178444 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
Mas Polyclonal specifically detects Mas in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Mas | |
Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:25, Immunohistochemistry-Paraffin 1:100 | |
Polyclonal | |
Rabbit | |
GPCR, Neuroscience | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
MAS, MAS1 oncogene, MGC119966, proto-oncogene Mas | |
MAS1 | |
IgG | |
Affinity Purified | |
Human, monkey, rat, and mouse. Rabbit reactivity reported in the scientific literature (PMID: 23701246). |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Primate, Rabbit | |
P04201 | |
4142 | |
A synthetic peptide made to an internal portion of the human MAS1 protein (between residues 75-125) [Uniprot: P04201] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title