Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mas Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP178444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178444 0.1 mL
NBP178444SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP178444 Supplier Novus Biologicals Supplier No. NBP178444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 13 publications

Mas Polyclonal specifically detects Mas in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Mas
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:25, Immunohistochemistry-Paraffin 1:100
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. P04201
Gene Alias MAS, MAS1 oncogene, MGC119966, proto-oncogene Mas
Gene Symbols MAS1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human MAS1 protein (between residues 75-125) [Uniprot: P04201]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4142
Test Specificity Human, monkey, rat, and mouse. Rabbit reactivity reported in the scientific literature (PMID: 23701246).
Target Species Human, Mouse, Rat, Primate, Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.