Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MDH1 Rabbit anti-Human, Polyclonal, Invitrogen™

Catalog No. PIPA535307
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535307 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535307 Supplier Thermo Scientific Supplier No. PA535307

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, chicken, mouse, porcine and rat based on sequence homology.

MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDH1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene MDH1
Gene Accession No. P40925
Gene Alias MDH1; MDHA; Malate dehydrogenase, cytoplasmic; Cytosolic malate dehydrogenase; Diiodophenylpyruvate reductase
Gene Symbols MDH1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 286-314 amino acids from the C-terminal region of human MDH1
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4190
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.