Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Microtubule-Associated Protein-2 (MAP2), Clone: AP20, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB3418X/DEL
Description
Immunohistochemistry, Western Blotting
Specifications
| Microtubule-Associated Protein-2 (MAP2) | |
| Alexa Fluor 488 | |
| Bovine brain microtubule protein. | |
| RUO | |
| Primary | |
| Semi-purified |
| Monoclonal | |
| Mouse | |
| 100 μg | |
| Neuroscience | |
| Bovine, Chicken, Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction