Learn More
MilliporeSigma™ anti-Microtubule-Associated Protein-2 (MAP2), Clone: AP20, ™ (Chemicon™)
Specifications
| Antigen | Microtubule-Associated Protein-2 (MAP2) |
|---|---|
| Classification | Monoclonal |
| Form | Semi-purified |
| Host Species | Mouse |
| Regulatory Status | RUO |
| Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Conjugate | Quantity | Price | Quantity & Availability | ||||
|
50-175-282
|
MilliporeSigma™
MAB3418X/DEL |
Alexa Fluor 488 | 100 μg | N/A | N/A | N/A | |||
|
50-175-281
|
MilliporeSigma™
MAB3418/DEL |
Unconjugated | 200 μg | N/A | N/A | N/A | |||
Description
Immunohistochemistry, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
| Microtubule-Associated Protein-2 (MAP2) | |
| Semi-purified | |
| RUO | |
| Bovine brain microtubule protein. |
| Monoclonal | |
| Mouse | |
| Neuroscience | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.