Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse ABCG2/CD338, Clone: BXP-21, Novus Biologicals™
SDP

Catalog No. NB1002177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002177 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002177 Supplier Novus Biologicals Supplier No. NB1002177

Mouse Monoclonal Antibody has been used in 7 publications

ABCG2/CD338 Monoclonal specifically detects ABCG2/CD338 in Human, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen ABCG2/CD338
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BXP-21
Concentration 0.25 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:20-1:50, Immunohistochemistry-Paraffin 1:20-1:40, Immunohistochemistry-Frozen 1:20-1:40
Gene Accession No. Q9UNQ0
Gene Alias ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter
Gene Symbols ABCG2
Host Species Mouse
Immunogen Fusion protein composed of E. Coli maltose binding protein and BCRP peptide [126 amino acids, 271-396 AA of BCRP.
Purification Method Tissue culture supernatant
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Hematopoietic Stem Cell Markers, Stem Cell Markers, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 9429
Test Specificity This recognizes an internal epitope of human BCRP (70 kDa), a transmembrane half-transporter. The does not cross-react with the human MDR1, MRP1, and MRP2 gene products.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.