Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-beta II Tubulin A, Clone: TUB 2.1, Novus Biologicals™
SDP

Catalog No. NB12011308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12011308 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12011308 Supplier Novus Biologicals Supplier No. NB12011308

Mouse Monoclonal Antibody

beta II Tubulin A Monoclonal specifically detects beta II Tubulin A in Human, Mouse, Rat, Amphibian, Bovine, Chicken, Hamster, Plant samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen beta II Tubulin A
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone TUB 2.1
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Simple Western, Immunocytochemistry/Immunofluorescence 1:200
Gene Accession No. P85108
Gene Alias dJ40E16.7, TUBB, TUBB2, TUBB2B, tubulin beta-2A chain, tubulin, beta 2, tubulin, beta 2A, tubulin, beta polypeptide, tubulin, beta polypeptide 2
Gene Symbols TUBB2A
Host Species Mouse
Immunogen Purified rat brain tubulin.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7280
Test Specificity Recognizes all five isoforms of beta-tubulin. It reacts with the beta-Lc and beta-Sc fragments in the carboxy-terminal part of beta-tubulin in immunoblotting.
Target Species Human, Mouse, Rat, Amphibian, Bovine, Chicken, Hamster, Plant
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.