Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse C-Reactive Protein/CRP, Clone: C3, Novus Biologicals™
SDP

Catalog No. NB200441 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200441 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200441 Supplier Novus Biologicals Supplier No. NB200441

Mouse Monoclonal Antibody

C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunoassay, Sandwich ELISA, Immunoaffinity Purification.

Specifications

Antigen C-Reactive Protein/CRP
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone C3
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Gene Alias C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895
Gene Symbols CRP
Host Species Mouse
Immunogen Full length native protein (purified) (Human).
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 1401
Test Specificity C Reactive Protein (C3)
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.