Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPR-Cas9 Antibody (7A9-3A3) - N-Terminus - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236440SS
Description
CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry Whole-Mount, Immunoassay.Specifications
| CRISPR-Cas9 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation, Immunohistochemistry-Frozen reported in scientific literature (PMID 28153089), Immunoblotting reported in scientific literature (PMID 31959836), Immunohistochemistry (Whole-Mount), Knockout Validated reported in scientific literature (PMID 31959836) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bacteria, Chicken, Fungi, Fungi | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoblot, Immunoassay | |
| 7A9-3A3 | |
| Unconjugated | |
| Cas9, CRISPR, CRISPR-associated protein 9 nuclease, SpyCas9 | |
| This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus was raised against Recombinant Cas9 within the N-terminal region of Streptococcus pyogene. [Uniprot: Q99ZW2]. | |
| 0.025 mL | |
| Gene Editing, Gene Knockouts, Gene Therapy | |
| Cas9 protein from Streptococcus pyogene serotype M1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction