Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD5, FITC, Clone: OX-19, Novus Biologicals™
SDP

Catalog No. NB10063914 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063914 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063914 Supplier Novus Biologicals Supplier No. NB10063914

Specifications

Antigen CD5
Applications Flow Cytometry
Classification Monoclonal
Clone OX-19
Concentration 0.1mg/mL
Conjugate FITC
Host Species Mouse
Immunogen Rat thymocyte glycoproteins
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 921
Test Specificity NB100-63914 recognizes the rat CD5 cell surface antigen, a 69kD glycoprotein expressed by T cells, thymocytes and a subset of B cells. OX-19 has been reported as being suitable for use on periodate-lysine paraformaldehyde (PLP) fixed paraffin embedded tissue (4). This product is routinely tested in flow cytometry on rat splenocytes.
Target Species Rat
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.