Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Clostridium botulinum B Toxoid, Clone: KBB18, Novus Biologicals™
SDP

Catalog No. NB10073117 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073117 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073117 Supplier Novus Biologicals Supplier No. NB10073117

Mouse Monoclonal Antibody

Clostridium Botulinum B Toxoid Monoclonal specifically detects Clostridium Botulinum B Toxoid in Bacteria samples. It is validated for Sandwich ELISA.

Specifications

Antigen Clostridium Botulinum B Toxoid
Applications ELISA
Classification Monoclonal
Clone KBB18
Conjugate Unconjugated
Dilution Sandwich ELISA 1:100-1:2000
Gene Alias BoNT, BoNT/B, BoNT/B Bontoxilysin B, Bontoxilysin B, BotB, Botulinum neurotoxin type B, Botulism, C botulinum, Clostridium botulinum
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with C. botulinum B toxoid.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity C. botulinum B toxoid. There is no cross-reactivity with A and E toxoids in sandwich ELISA.
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.