Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-COL8A2, Clone: V-3C9, Novus Biologicals™
SDP

Catalog No. NBP229851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229851 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229851 Supplier Novus Biologicals Supplier No. NBP229851

Specifications

Antigen COL8A2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone V-3C9
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5-10ug/ml, ELISA 1:100-1:2000, Immunohistochemistry-Paraffin 1-10 ug/ml
Gene Accession No. P20908
Gene Alias alpha-2 polypeptide, collagen, type VIII, alpha 2, dJ665N4.1 (collagen type VIII alpha 2), FECD, FLJ00201, MGC116970, PPCD, PPCD2MGC116972
Gene Symbols COL8A2
Host Species Mouse
Immunogen This antibody is specific to Collagen Type V
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1296
Test Specificity The antibody specifically reacts with the hCL(V). This is a purified mouse monoclonal antibody to human type V collagen (hCL(V)). SPECIES REACTIVITY: This antibody cross-reacts with rabbit and rat CL(V).
Target Species Human, Rat, Rabbit
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.