Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CTRP2, Clone: Didoo-1, Novus Biologicals™
SDP

Catalog No. NBP197580 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP197580 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP197580 Supplier Novus Biologicals Supplier No. NBP197580

Mouse Monoclonal Antibody

CTRP2 Monoclonal specifically detects CTRP2 in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen CTRP2
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone Didoo-1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:200
Gene Alias C1q and tumor necrosis factor related protein 2, complement-c1q tumor necrosis factor-related protein 2, CTRP2complement C1q tumor necrosis factor-related protein 2, zacrp2
Gene Symbols C1QTNF2
Host Species Mouse
Immunogen Recombinant mouse CTRP2 (C1q tumor necrosis factor-alpha-related protein 2) (aa 26-260).
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114898
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.