Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse Cystatin E/M/CST6, PE, Clone: MM0231-4H54, Novus Biologicals™
SDP

Catalog No. NBP212251PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212251PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212251PE Supplier Novus Biologicals Supplier No. NBP212251PE

Mouse Monoclonal Antibody

Cystatin E/M/CST6 Monoclonal specifically detects Cystatin E/M/CST6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Cystatin E/M/CST6
Applications Flow Cytometry
Classification Monoclonal
Clone MM0231-4H54
Conjugate PE
Dilution Flow Cytometry
Gene Accession No. Q15828
Gene Alias cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor
Gene Symbols CST6
Host Species Mouse
Immunogen Human recombinant Cystatin E/M/CST6
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1474
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.