Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Glycogen Phosphorylase BB/GPBB, Clone: 3G1, Novus Biologicals™
SDP

Catalog No. NB120882 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120882 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120882 Supplier Novus Biologicals Supplier No. NBP244176JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GSTO1 Monoclonal specifically detects GSTO1 in Human, Mouse samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen Glycogen Phosphorylase BB/GPBB
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3G1
Conjugate Unconjugated
Host Species Mouse
Immunogen Full length protein (Human).
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5834
Test Specificity Reacts with the BB and MM isoenzyme. Cross-reacts with Human. Not yet tested in other species.
Target Species Human
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.