Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GM-CSF, Clone: 30-4, Novus Biologicals™
SDP

Catalog No. NB10065022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065022 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065022 Supplier Novus Biologicals Supplier No. NB10065022

Mouse Monoclonal Antibody

GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional.

Specifications

Antigen GM-CSF
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay
Classification Monoclonal
Clone 30-4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA 0.5-1 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 8-25 ug/ml, Functional 0.3-0.5 ug/ml
Gene Accession No. P04141
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Gene Symbols CSF2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 14 to 144 of human GM-CSF produced in E. coli.
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 1437
Test Specificity NB100-65022 recognizes recombinant and native human granulocyte macrophage colony stimulating factor (GM-CSF). Human Granulocyte colony stimulating factor (GM-CSF) is a 14.6kDa glycoprotein, which is produced by T cells, macrophages, fibroblasts and endothelial cells. GM-CSF is involved in the stimulation of proliferation and differentiation of granulocyte and macrophage progenitor cells. The neutralisation dose (ND50) of this batch, as determined by the proliferation of TF-1 cells, is approximately 0.3-0.5 ug/ml in the presence of 0.5 ng/ml of recombinant human GM-CSF. The ND50 may vary depending on conditions.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.