Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IGF-I R, Clone: 3B7, Novus Biologicals™
SDP

Catalog No. NB120985 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120985 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120985 Supplier Novus Biologicals Supplier No. NBP286683JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

c-Myc Monoclonal specifically detects c-Myc in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen IGF-I R
Applications Immunoprecipitation
Classification Monoclonal
Clone 3B7
Concentration 1.1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified human placental IGF-1 receptors.
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3480
Test Specificity This reacts specifically with the IGF-1 receptor. This stimulates binding of IGF-1 and -2 to the receptor and stimulates basal and IGF-1 stimulated cell growth. Cross-reacts with Human. Not yet tested in other species.
Target Species Human
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.