Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IL-19, PE, Clone: MM0383-1G54, Novus Biologicals™
SDP

Catalog No. NBP211681PE Shop All R&D Systems Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211681PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. NBP211681PE missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP211681PE

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

IL-19 Monoclonal antibody specifically detects IL-19 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen IL-19
Applications Flow Cytometry
Classification Monoclonal
Clone MM0383-1G54
Conjugate PE
Dilution Flow Cytometry
Gene Accession No. AAN40906
Gene Alias IL-10C, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG.1Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19
Gene Symbols IL19
Host Species Mouse
Immunogen Produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human recombinant protein of IL-19.
Purification Method >95% pure by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 29949
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.