Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A Hemagglutinin H5 Mouse anti-Virus, Clone: 19C11, Novus Biologicals™
SDP

Catalog No. NBP105463 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105463 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105463 Supplier Novus Biologicals Supplier No. NBP105463

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Influenza A Hemagglutinin H5 Monoclonal antibody specifically detects Influenza A Hemagglutinin H5 in Virus samples. It is validated for Immunoassay, Hemagglutination Assay, Dot Blot, ELISA.

Specifications

Antigen Influenza A Hemagglutinin H5
Applications Immunoassay, Hemagglutination Assay, Dot Blot, ELISA
Classification Monoclonal
Clone 19C11
Conjugate Unconjugated
Dilution Immunoassay, Haemagglutination, Dot Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000
Gene Alias H5HA, Hemagglutinin 5
Host Species Mouse
Immunogen react with hemaglutinin H5 in hemagluttinin inhibition (HI) test
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity react with hemaglutinin H5 in hemagluttinin inhibition (HI) test
Target Species Avian
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.