Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Insulin, Clone: 3A6, Novus Biologicals™
SDP

Catalog No. NB10073008 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073008 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073008 Supplier Novus Biologicals Supplier No. NB10073008

Mouse Monoclonal Antibody has been used in 2 publications

Insulin Monoclonal specifically detects Insulin in Human, Mouse samples. It is validated for ELISA, Enzyme Immunoassay, Sandwich ELISA.

Specifications

Antigen Insulin
Applications ELISA, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 3A6
Conjugate Unconjugated
Dilution ELISA, Enzyme Immunoassay, Sandwich ELISA 1:100-1:2000
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Gene Symbols INS
Host Species Mouse
Immunogen Derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with purified human insulin.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Diabetes Research, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3630
Test Specificity This cross-reacts with human proinsulin, bovine insulin (30%) and porcine insulin. No cross-reaction with free C-peptide.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.