Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Marburg Virus, Clone: FM11, Novus Biologicals™
SDP

Catalog No. NB1107402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1107402 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1107402 Supplier Novus Biologicals Supplier No. NB1107402

Mouse Monoclonal Antibody

Marburg Virus Monoclonal specifically detects Marburg Virus in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Marburg Virus
Applications Western Blot, ELISA
Classification Monoclonal
Clone FM11
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Gene Alias MBG
Host Species Mouse
Immunogen Marburg virus.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Marburg Virus (FM11)
Target Species Virus
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.