Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mycobacterium tuberculosis 38kDa, Clone: HTM81, Novus Biologicals™
SDP

Catalog No. NB10073190 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073190 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073190 Supplier Novus Biologicals Supplier No. NB10073190

Mouse Monoclonal Antibody

Mycobacterium tuberculosis 38kDa Monoclonal specifically detects Mycobacterium tuberculosis 38kDa in Bacteria samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Mycobacterium tuberculosis 38kDa
Applications Western Blot, ELISA
Classification Monoclonal
Clone HTM81
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Host Species Mouse
Immunogen M. tuberculosis 38 kDa antigen
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity M. tuberculosis 38 kDa antigen
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.