Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Myoglobin, Clone: 7C3, Novus Biologicals™
SDP

Catalog No. NB1208345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1208345 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1208345 Supplier Novus Biologicals Supplier No. NB1208345

Mouse Monoclonal Antibody

Myoglobin Monoclonal specifically detects Myoglobin in Human samples. It is validated for Sandwich ELISA.

Specifications

Antigen Myoglobin
Applications ELISA
Classification Monoclonal
Clone 7C3
Conjugate Unconjugated
Dilution Sandwich ELISA 1:100-1:2000
Gene Accession No. P02144
Gene Alias MGC13548, myoglobin, PVALB
Gene Symbols MB
Host Species Mouse
Immunogen Human cardiac myoglobin
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 4151
Test Specificity Human cardiac myoglobin
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.