Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rabies Virus, Clone: 4F1, Novus Biologicals™
SDP

Catalog No. NBP211627 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211627 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211627 Supplier Novus Biologicals Supplier No. NBP211627

Mouse Monoclonal Antibody

Rabies Virus Monoclonal specifically detects Rabies Virus in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence.

Specifications

Antigen Rabies Virus
Applications ELISA
Classification Monoclonal
Clone 4F1
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Gene Alias Glycoprotein
Host Species Mouse
Immunogen specific to glycoprotein
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.