Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Renal Cell Carcinoma (gp200), APC, Clone: HD30, Novus Biologicals™
SDP

Catalog No. NBP141003J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141003J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141003J Supplier Novus Biologicals Supplier No. NBP141003APC

Mouse Monoclonal Antibody

DEC-205/CD205 Monoclonal specifically detects DEC-205/CD205 in Human, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen Renal Cell Carcinoma (gp200)
Applications Flow Cytometry
Classification Monoclonal
Clone HD30
Conjugate APC
Dilution Flow Cytometry
Gene Accession No. O60449
Gene Alias CD205LY-75, CLEC13BLy-75, C-type lectin domain family 13 member B, DEC-205CD205 antigen, GP200-MR6, lymphocyte antigen 75
Gene Symbols LY75
Host Species Mouse
Immunogen Purified extracellular domain of human CD205-IgG1 Fc fusion protein.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers, Vision
Primary or Secondary Primary
Gene ID (Entrez) 4065
Test Specificity This antibody specifically recognises CD205, otherwise known as DEC205, a type I transmembrane glycoprotein and member of the macrophage mannose receptor family of C-type lectins, predominantly expressed by human dendritic cells.
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.