Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rotavirus Inner Capsid Protein, Clone: 2B4, Novus Biologicals™
SDP

Catalog No. NB11037240 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11037240 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11037240 Supplier Novus Biologicals Supplier No. NB11037240

Specifications

Antigen Rotavirus Inner Capsid Protein
Applications Western Blot, ELISA, Immunoassay
Classification Monoclonal
Clone 2B4
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1:1000, ELISA 1:2000-1:10000, Immunoassay 1:100-1:500
Formulation Tris 0,1M, glycine 0,1M, sucrose 2% with No Preservative
Host Species Mouse
Immunogen Recombinant inner capsid protein (VP6).
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute to desired concentration.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.