Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SLC6A2/NET/Noradrenaline transporter, Clone: NET-05, Novus Biologicals™
SDP

Catalog No. NBP128665 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128665 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128665 Supplier Novus Biologicals Supplier No. NBP128665

Specifications

Antigen SLC6A2/NET/Noradrenaline transporter
Applications Western Blot
Classification Monoclonal
Clone NET-05
Conjugate Unconjugated
Dilution Western Blot 1:2000
Gene Accession No. O55192
Gene Alias NAT1neurotransmitter transporter, NET, NET1sodium-dependent noradrenaline transporter, Norepinephrine transporter, SLC6A5, solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2, Solute carrier family 6 member 2, solute carrier family 6 member 5
Gene Symbols SLC6A2
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acid residues from the N-terminal region conjugated to KLH
Molecular Weight of Antigen 55 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6530
Test Specificity Specific for the ∼55kDa Norepinephrine Transporter in Western blots of mouse and rat cortex homogenate. No reactivity with tissue or cells from NET knock-out mice.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.