Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRO-1 Antibody (STRO-1), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 8 publications

$206.50 - $499.50

Specifications

Antigen STRO-1
Clone STRO-1
Concentration 1.0 mg/mL
Dilution Flow Cytometry 1:400, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunocytochemistry
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP148356SS
SDP
View Documents
Novus Biologicals
NBP148356SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP148356
SDP
View Documents
Novus Biologicals
NBP148356
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

STRO-1 Monoclonal specifically detects STRO-1 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunocytochemistry.
Specifications

Specifications

STRO-1
1.0 mg/mL
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Stem Cell Markers
STRO1
IgM λ
IgM purified
STRO-1
Flow Cytometry 1:400, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunocytochemistry
Monoclonal
Purified
RUO
Human, Rat
Human CD34+ bone marrow cells.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.