Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRO-1 Antibody (STRO-1), Novus Biologicals™
SDP

Catalog No. NBP148356SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP148356SS 0.025 mL
NBP148356 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP148356SS Supplier Novus Biologicals Supplier No. NBP148356SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 8 publications

STRO-1 Monoclonal specifically detects STRO-1 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunocytochemistry.

Specifications

Antigen STRO-1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone STRO-1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:400, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunocytochemistry
Gene Alias STRO1
Host Species Mouse
Immunogen Human CD34+ bone marrow cells.
Purification Method IgM purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.