Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TCR gamma/delta, FITC, Clone: TCR1, Novus Biologicals™
SDP

Catalog No. NBP128276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128276 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128276 Supplier Novus Biologicals Supplier No. NBP128276

Specifications

Antigen TCR gamma/delta
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone TCR1
Concentration 0.5mg/mL
Conjugate FITC
Dilution Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay
Gene Alias T cell receptor gamma locus, T-cell antigen receptor, gamma polypeptide, T-cell rearranging gene, gamma, T-cell receptor, gamma cluster, TCRG, TRG
Gene Symbols TRG
Host Species Mouse
Immunogen Outbred chicken thymocytes and Ig-negative blood lymphocytes
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6965
Target Species Chicken, Reptile
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.