Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-XRCC1, Clone: 33-2-5, Novus Biologicals™
SDP

Catalog No. NBP226439 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226439 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226439 Supplier Novus Biologicals Supplier No. NBP226439

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen XRCC1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 33-2-5
Conjugate Unconjugated
Host Species Mouse
Immunogen BALB/C mice were injected with recombinant human XRCC1 protein.
Purification Method Affinity Purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7515
Target Species Rat
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.