Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Yersinia pestis V Antigen, Clone: Va13, Novus Biologicals™
SDP

Catalog No. NB10073227 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073227 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073227 Supplier Novus Biologicals Supplier No. NB10073227

Mouse Monoclonal Antibody

Yersinia Pestis V Antigen Monoclonal specifically detects Yersinia Pestis V Antigen in Bacteria samples. It is validated for Western Blot, Sandwich ELISA.

Specifications

Antigen Yersinia Pestis V Antigen
Applications Western Blot, ELISA
Classification Monoclonal
Clone Va13
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000
Gene Alias LcrV, Low calcium response locus protein V, V antigen antihost protein/regulator, V antigen LcrV, Virulence associated V antigen
Host Species Mouse
Immunogen Y. pestis, V antigen
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Y. pestis, V antigen
Target Species Bacteria
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.