Learn More
Munc-18 Mouse, Unlabeled, Clone: 31, BD
Mouse Monoclonal Antibody
$287.50 - $568.50
Specifications
| Antigen | Munc-18 |
|---|---|
| Clone | 31 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
There are two Munc-18 isoforms: Munc-18-1 and Munc-18-2. Munc-18-1 (n-sec1 or rb-sec1; 68 kDa) is a neuronal protein that binds tightly to syntaxin 1 and functions in synaptic vesicle exocytosis. Munc-18-2 exhibits 63% amino acid sequence identity with Munc-18-1. Although Munc-18-2 is expressed in most tissues, Munc-18-1 is primarily expressed in brain. Both forms of Munc-18 bind tightly to syntaxins 1A, 2, and 3, but not to syntaxin 4. Syntaxin 1A is a plasma membrane protein implicated in synaptic vesicle docking. Following calcium entry into the presynaptic nerve terminal, the neurotransmitter-containing synaptic vesicle fuses with the plasma membrane at a region known as the active zone. Thus, by virtue of its interaction with syntaxin 1A, Munc-18-1 is thought to have an essential function in neurotransmitter release.
Specifications
| Munc-18 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 31 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.