Learn More
NCK Mouse, Unlabeled, Clone: 108, BD
Mouse Monoclonal Antibody
$286.00
Specifications
Antigen | NCK |
---|---|
Clone | 108 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
NCK is a 47kDa protein that belongs to a class of signaling molecules called adaptors. These proteins exert their effects by coupling activated receptors to downstream effectors. NCK is comprised almost entirely of one SH2 and three SH3 domains and it possesses no intrinsic catalytic activity. Investigations of its interactions with cellular proteins have found constitutive association of NCK with various Ser/THr kinases, such as p21-associated kinase (PAK) and casein kinase 1. Other proteins that directly interact with NCK are: Sos, Cbl, Wiskott-Aldrich syndrome protein (WASP), IRS1, and multiple tyrosine kinase receptors. The presence of NCK is necessary for the mitogenic response elicited by PDGF in some cells. In addition, the Drosophila homologue of NCK is involved in regulating cytoskeletal organization.
Host Species: Mouse
Clone: 108
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Human NCK aa. 279-377
Formula Weight [Chemical]: 47kDa
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
NCK | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2b | |
Affinity Purified |
108 | |
Western Blot | |
Unconjugated | |
RUO | |
Bovine, Canine, Chicken, Frog, Human, Mouse, Rat | |
Human NCK aa. 279-377 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.