Learn More
NeuN Mouse anti-Mouse, Rat, Human, Ferret, Chicken, Salamander, Biotin, Clone: A60, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | NeuN |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | A60 |
| Conjugate | Biotin |
| Formulation | Purified mouse monoclonal IgG1 in buffer containing 0.01MPBS pH 7.1, 0.1% sodium azide with 15mg/mL BSA as a stabilizer. |
| Gene Symbols | NeuN, A59 |
| Host Species | Mouse |
| Immunogen | Purified cell nuclei from mouse brain |
| Purification Method | Protein A purfied |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.