Learn More
Neurotensin Receptor 3 (NTR3) Mouse, Unlabeled, Clone: 48, BD
Mouse Monoclonal Antibody
$286.00 - $566.00
Specifications
Antigen | Neurotensin Receptor 3 (NTR3) |
---|---|
Clone | 48 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Neurotensin (NT) is a neuropeptide that modulates dopaminergic transmission, triggers analgesic responses, induces hypotension, decreases gastric acid secretion, and activates lipid digestion. NT is a ligand for NT receptors, which include both G-protein coupled and non-G-protein coupled receptors. Neurotensin Receptor 3 (NTR3) is a non-G-protein coupled receptor that has a lumenal domain homologous to sorting proteins, and a short cytoplasmic tail homologous to mannose-6-phosphate/IGF-II receptor. NTR3 was also identified as sortilin/gp95, a component of GLUT4 vesicles in adipocytes that has been implicated in lipoprotein lipase degradation. NTR3 mRNA is expressed in brain, skeletal muscle, heart, and adipocytes. NTR3 mature protein has 44 N-terminal amino acid residues cleaved off, which may facilitate ligand binding to the receptor. Cellular localization of NTR3 is in the Golgi compartment and vesicles, as well as on the cell surface. In addition to roles in neuropeptide and lipoprotein degradation, NTR3 may also be involved in receptor protein sorting. Thus, NTR3 is a multifunctional transmembrane protein that acts as both a intracellular sorting receptor and extracellular ligand-binding receptor.
Immunofluorescence, Western Blotting
Specifications
Neurotensin Receptor 3 (NTR3) | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
NTR3 |
48 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Human Neurotensin Receptor 3 aa. 300-422 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.