Learn More
NMDAR2B Mouse, Unlabeled, Clone: 13, BD
Mouse Monoclonal Antibody
$286.00 - $566.00
Specifications
Antigen | NMDAR2B |
---|---|
Clone | 13 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The majority of synapses in the central nervous system utilize glutamate as a neurotransmitter to produce rapid neuronal excitation. Glutamate has a diverse array of receptors that can be categorized into two groups: ionotropic and metabotropic. The ionotropic receptors are subdivided into two distint types: 1) receptors for N-methyl D-aspartate (NMDAR) and 2) non-NMDA receptors for AMPA and kainate. Three types of NMDAR2 have been identified: NR2A, NR2B, and NR2C. NR2A and NR2B contain a C-terminal extension (>600 amino acids) that has small scattered regions of conserved sequence. The three NR2 mRNAs show overlapping, differential expression patterns in the rat brain. NR2B has been reported to be expressed in the forebrain, thalamic nuclei, amygdaloid nuclei, caudateputamen, and in restricted regions of the olfactory bulb.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
NMDAR2B | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2b | |
Affinity Purified | |
N-Methyl-D-Aspartate Receptor 2B |
13 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.