Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nNOS Mouse, Unlabeled, Clone: 16, BD

Catalog No. BDB610308 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610308 50 μg
BDB610309 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610308 Supplier BD Biosciences Supplier No. 610308
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nitric oxide synthase (NOS), a cell-type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits cellular signals involved in vasorelaxation, neurotransmission, and cytotoxicity. In neurons and endothelial cells, constitutive NOS (cNOS) is activated by agonists that increase intracellular Ca2+ levels and enhance calmodulin binding. Neuronal NOS (nNOS or bNOS) and endothelial NOS (ECNOS) have recognition sites for NADPH, FAD, FMN, and calmodulin and are regulated in a similar manner. However, both have been shown to be distinct gene products of about 155kDa and 140kDa, respectively, and the human forms show 52% amino acid identity. Neuronal NOS and induced macrophage NOS (iNOS) share 51% amino acid homology with the greatest degree of divergence in the calmodulin binding domain. Neuronal NOS, a cytosolic protein present mainly in neural tissues, has been purified and characterized from rat cerebellum. The NO synthesized by this enzyme acts as a neurotransmitter. ECNOS has been cloned from human vascular endothelium as well as from bovine aortic endothelial cells (BAEC) and has a unique N-myristylation consensus sequence that may explain its membrane localization.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen nNOS
Applications Western Blot
Classification Monoclonal
Clone 16
Concentration 250μg/mL
Conjugate Unconjugated
Description NOS Type I; Neuronal Nitric Oxide Synthase
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.