Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-NSD3, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP104942 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104942 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104942 Supplier Novus Biologicals Supplier No. NBP104942

Rabbit Polyclonal Antibody

NSD3 Polyclonal specifically detects NSD3 in Human samples. It is validated for Western Blot, ICC/IF (Negative).

Specifications

Antigen NSD3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ICC/IF (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
Gene Accession No. Q9BZ95
Gene Alias EC 2.1.1.43, FLJ20353, histone-lysine N-methyltransferase NSD3, MGC126766, MGC142029, NSD3DKFZp667H044, Nuclear SET domain-containing protein 3, pp14328, Protein whistle, WHSC1-like 1 isoform 9 with methyltransferase activity to lysine, WHSC1-like protein 1, Wolf-Hirschhorn syndrome candidate 1-like 1, Wolf-Hirschhorn syndrome candidate 1-like protein 1
Gene Symbols WHSC1L1
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human NSD3 protein (within residues 300-400). [Swiss-Prot# Q9BZ95]
Molecular Weight of Antigen 165 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 54904
Test Specificity This has been observed to recognize a band that corresponds to isoform 1 of NSD3.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.