Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleolin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 23 publications
$206.50 - $499.50
Specifications
| Antigen | Nucleolin |
|---|---|
| Dilution | Western Blot 1:4000, Simple Western 1:400, Chromatin Immunoprecipitation reported in scientific literature, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 30411850), Immunomicroscopy, Electron Microscopy 1:10-1:500, Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 22483616), Chromatin Immunoprecipitation (ChIP), KnockDown Validated |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB600241SS
![]() |
Novus Biologicals
NB600241SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB600241
![]() |
Novus Biologicals
NB600241 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Nucleolin Polyclonal specifically detects Nucleolin in Human, Mouse, Rat, Plant samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunomicroscopy, Electron Microscopy, Immunohistochemistry Whole-Mount, Knockdown Validated.Specifications
| Nucleolin | |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, DNA replication Transcription Translation and Splicing | |
| Whole antisera with 0.1% Sodium Azide | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| NCL | |
| IgG | |
| Unpurified | |
| Human, mouse, rat and soy bean plant. |
| Western Blot 1:4000, Simple Western 1:400, Chromatin Immunoprecipitation reported in scientific literature, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 30411850), Immunomicroscopy, Electron Microscopy 1:10-1:500, Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 22483616), Chromatin Immunoprecipitation (ChIP), KnockDown Validated | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Plant | |
| P19338 | |
| 4691 | |
| A fusion protein containing amino acids 284-709 of human Nucleolin. [UniProt# P19338] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 106 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title