Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleolin Antibody, Novus Biologicals™
SDP

Catalog No. NB600241 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600241 0.1 mL
NB600241SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600241 Supplier Novus Biologicals Supplier No. NB600241
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 23 publications

Nucleolin Polyclonal specifically detects Nucleolin in Human, Mouse, Rat, Plant samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunomicroscopy, Electron Microscopy, Immunohistochemistry Whole-Mount, Knockdown Validated.

Specifications

Antigen Nucleolin
Applications Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:4000, Simple Western 1:400, Chromatin Immunoprecipitation reported in scientific literature, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen reported in scientific literature (PMID 30411850), Immunomicroscopy, Electron Microscopy 1:10-1:500, Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 22483616), Chromatin Immunoprecipitation (ChIP), KnockDown Validated
Formulation Whole antisera with 0.1% Sodium Azide
Gene Accession No. P19338
Gene Alias C23, FLJ45706, FLJ59041, nucleolin, Protein C23
Gene Symbols NCL
Host Species Rabbit
Immunogen A fusion protein containing amino acids 284-709 of human Nucleolin. [UniProt# P19338]
Molecular Weight of Antigen 106 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 4691
Test Specificity Human, mouse, rat and soy bean plant.
Target Species Human, Mouse, Rat, Plant
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.