Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human OCIL/CLEC2d PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3480P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3480P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3480P Supplier R&D Systems Supplier No. FAB3480P
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

OCIL/CLEC2d Monoclonal specifically detects OCIL/CLEC2d in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen OCIL/CLEC2d
Applications Flow Cytometry
Classification Monoclonal
Clone 402659
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9UHP7
Gene Alias CLAXLLT-1, CLEC2D, C-type lectin domain family 2 member D, C-type lectin domain family 2, member D, C-type lectin related f, Lectin-like NK cell receptor, Lectin-like transcript 1, LLT1, LLT1Osteoclast inhibitory lectin, OCILmember D
Gene Symbols CLEC2D
Host Species Mouse
Immunogen E. coli-derived recombinant human OCIL/CLEC2d Ser57-Val191 Accession # Q9UHP7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29121
Test Specificity Detects human OCIL/CLEC2d.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.