Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-OX40L (CD252), PE-Cy7, Clone: RM134L,
Rat Monoclonal Antibody
Supplier: MilliporeSigma™ MABF1446
Description
Specifically detects OX40L (CD252) Clone: RM134L in Mouse samples, and it is validated for Flow Cytometry
The RM134L antibody recognizes CD252, also known as OX-40 Ligand or CD134 Ligand, a member of the TNF superfamily that is present on the surface of antigen presenting cells and activated B lymphocytes. The OX-40 Ligand interacts with OX-40 (CD134) which is expressed primarily on activated T cells. This costimulatory interaction leads to increased proliferation and IL-2 production responses of activated T cells, and at the same time enhances proliferation and immunoglobulin secretion by activated B cells.The RM134L antibody is useful for flow cytometric detection of CD252 on stimulated mouse splenocytes.
Specifications
OX40L (CD252) | |
Monoclonal | |
PE-Cyanine7 | |
P43488 | |
Rat | |
Purified by size-exclusion chromatography | |
RUO | |
Primary | |
2°C to 8°C for one year from date of shipment protected from light; Do not freeze | |
IgG2b κ |
Flow Cytometry | |
RM134L | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Tnfsf4, Ox40l, Txgp1l | |
Proprietary immunogen | |
25 μg | |
Inflammation, Immunology | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction