Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p190-B Mouse, Unlabeled, Clone: 54, BD

Catalog No. p-3850311 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611612 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611612 Supplier BD Biosciences Supplier No. 611612

Mouse Monoclonal Antibody

GTPase activating proteins (GAPs) stimulate the GTP-hydrolyzing activity of GTPases, such as p21ras and Rho. p190-A is a Ras-GAP associated protein that is tyrosine phosphorylated in transformed and growth factor-stimulated cells. Ras-GAP and p190-A are targets of oncoproteins and growth factor receptors. p190-B is another Ras-GAP in the p190 family. It has 51% identity with p190-A and contains several GTPase-related domains in the N-terminal region and a Rho-GAP domain in the C-terminal region. p190-B is expressed in kidney, brain, liver, and lung, as well as in human foreskin fibroblasts, RD muscle cells, and HT-1080 cells. In fibroblasts, p190-B is localized diffusely in the cytoplasm and co-localizes with the α5β1 integrin receptor for fibronectin. Adhesion of fibronectin-coated latex beads to cells leads to the recruitment of p190-B and Rho to the plasma membrane at sites of bead contact. In addition, the recombinant Rho-GAP domain of p190-B displays GAP activity for RhoA, Rac1, and G25K/CDC42Hs. Thus, p190-B is thought to act as a transmembrane link between integrins and Rho GTPases during fibronectin-induced changes in cell morphology and motility.

Host Species: Mouse
Clone: 54
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human p190-B aa. 1102-1214

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen p190-B
Applications Western Blot
Classification Monoclonal
Clone 54
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human p190-B aa. 1102-1214
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.