Learn More
p230 trans Golgi Mouse anti-Human, Unlabeled, Clone: 15, BD
Mouse Monoclonal Antibody
$287.50 - $574.50
Specifications
Antigen | p230 trans Golgi |
---|---|
Clone | 15 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The Golgi apparatus is a very complex and dynamic organelle that functions in protein sorting and modification. Numerous structural and regulatory proteins are involved in the budding, docking, and fusion of Golgi-directed vesicles. p230 trans Golgi is a peripheral membrane protein associated with the cytosolic face of the trans Golgi network (TGN). Protein sequence analysis indicates that it is highly hydrophilic and contains two proline-rich regions, as well as a high frequency of heptad repeats which are characteristic of α-helices that form dimeric coiled-coiled structures. Additionally, p230 trans Golgi contains a ESLALEELEL sequence, a motif of the acidic granin proteins that are found in secretory granules of neuroendocrine cells. p230 trans Golgi cycles between the cytosol and the TGN via non-clathrin coated vesicles in a G protein-dependent manner. The localization of p230 trans Golgi to the TGN is dependent on aromatic residues in its C-terminal non-coiled coil domain. Thus, p230 trans-Golgi is aTGN-specific protein that is important for the biogenesis of distinct populations of non-clathrin coated vesicles and, thus, functions in vesicular transport from the TGN.
Host Species: Mouse
Clone: 15
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human p230 trans Golgi aa. 2063-2179
Formula Weight [Chemical]: 230kDa
Immunofluorescence, Western Blotting
Specifications
p230 trans Golgi | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
15 | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Human p230 trans Golgi aa. 2063-2179 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.