Learn More
p27[Kip1] Mouse, Unlabeled, Clone: 57, BD
Mouse Monoclonal Antibody
$287.50 - $568.50
Specifications
Antigen | p27[Kip1] |
---|---|
Clone | 57 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Kip1 is a cyclin-dependent kinase (cdk) inhibitor that was identified as a result of its role in TGFβ-induced G1 phase arrest and cell-cell contact. In vitro , p27 [Kip1] binds tightly to Cyclin D-Cdk4, Cyclin E-Cdk2, and Cyclin A-Cdk2 complexes and inhibits their activity. In normal cells, Kip1 is sequestered and its activity gradually decreases as cells reach S phase. The addition of TGF-β in early G1 blocks this decline in activity, by preventing the sequestration of Kip1. TGF-β also reduces the levels of Cdk4. Kip1 preferentially binds Cyclin D-Cdk4, but the lower levels of Cdk4 in TGF-β treated cells allow Kip1 to be available for binding to Cyclin E-Cdk2 and Cyclin A-Cdk2. Kip1 is structurally related to Cip1/WAF1, having a similar 60 amino acid sequence in the N-terminal region. A 52 amino acid Kip1 peptide (residues 28-79) from this region is sufficient to inhibit cdk activity in vitro.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
p27[Kip1] | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
57 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Frog, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.