Learn More
p67 (phox) Mouse, Unlabeled, Clone: 29, BD
Mouse Monoclonal Antibody
$286.00 - $528.50
Specifications
Antigen | p67 (phox) |
---|---|
Clone | 29 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The neutrophil respiratory burst oxidase (NADPH-oxidase) generates superoxide and secondary oxygen-derived toxic products in response to bacteria or a variety of soluble stimuli. The active site of this enzyme is located in an integral membrane cytochrome, b558, that consists of the two subunits gp91 [phox] and p21 [phox]. Superoxide production depends on the formation of a complex that includes p67 [phox], p47 [phox], and the GTP-binding protein Rac. Upon activation, these proteins translocate from the cytosol to the membrane where they assemble with b558 and induce oxidase activity. p67 [phox] contains two SH3 domains and binds, via its C-terminal SH3 domain, to the proline rich
region of p47 [phox]. This binding allows p67 [phox] to indirectly associate with the oxidase. It is thought that the phosphorylated forms of p67 [phox] and p47 [phox] interact and that the phosphorylation of p67 [phox] is regulated by both PKC-dependent and independent pathways. Although the role of p67 [phox] in electron flow control is poorly understood, it is thought that it regulates the transfer of electrons from NADPH to reduce flavin.
Immunofluorescence, Western Blotting
Specifications
p67 (phox) | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2b | |
Affinity Purified |
29 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse | |
Human p67 [phox] aa. 317-469 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.