Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Parkinson disease (autosomal recessive, early onset) 7 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004542 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-542 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-542 Supplier Abnova Corporation Supplier No. H00011315A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant PARK7.

The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq

Sequence: MASKRALVILAKGAEEMETVIPVDVMRRAGIKVTVAGLAGKDPVQCSRDVVICPDASLEDAKKEGPYDVVVLPGGNLGAQNLSESAAVKEILKEQENRKG

Specifications

Antigen Parkinson disease (autosomal recessive, early onset) 7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant PARK7.
Formulation 50% glycerol
Gene PARK7
Gene Accession No. BC008188
Gene Alias DJ-1/DJ1/FLJ27376/FLJ34360/FLJ92274
Gene Symbols PARK7
Host Species Mouse
Immunogen PARK7 (AAH08188, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 11315
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.