Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Pax5/BSAP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP225248 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225248 0.1 mg
NBP225248SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225248 Supplier Novus Biologicals Supplier No. NBP225248

Rabbit Polyclonal Antibody

Pax5/BSAP Polyclonal antibody specifically detects Pax5/BSAP in Human samples. It is validated for Western Blot.

Specifications

Antigen Pax5/BSAP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. NP_057306
Gene Alias B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5
Gene Symbols PAX5
Host Species Rabbit
Immunogen Within the range of amino acids 44-159 of human DDX41 were used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5079
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.